Stampa…
Cartridge filters, Impact® 2 V2
Fornitore: ENTEGRIS
A2AZ200K1 A2AG200K1 A2AV200K1 A2NT200K1 A2NX200K1 A2UA200K1 A2AG20001 A2AT20M01 A2AT20MK1 A2AV20001 A2AX20M01 A2AX20MK1 A2AZ20001 A2MX20M01 A2MX20MK1 A2NT20001 A2NX20001 A2SF20M01 A2SF20MK1 A2ST20M01 A2ST20MK1
ENTEA2AZ200K1EA 0 EUR
ENTEA2AZ200K1 ENTEA2AG200K1 ENTEA2AV200K1 ENTEA2NT200K1 ENTEA2NX200K1 ENTEA2UA200K1 ENTEA2AG20001 ENTEA2AT20M01 ENTEA2AT20MK1 ENTEA2AV20001 ENTEA2AX20M01 ENTEA2AX20MK1 ENTEA2AZ20001 ENTEA2MX20M01 ENTEA2MX20MK1 ENTEA2NT20001 ENTEA2NX20001 ENTEA2SF20M01 ENTEA2SF20MK1 ENTEA2ST20M01 ENTEA2ST20MK1
Cartridge filters, Impact® 2 V2
Filtri Cartucce con filtro
Superior retention performance for ultra-clean, point-of-use filtration of photoresist and photochemical solvents.
  • Spontaneously wets in butyl acetate, PGMEA, PGME, ethyl lactate, MMP, cyclohexanone, ECA, NMP and other photochemical solvents
  • Optimised for POU filtration of photoresist and photochemical solvents and aqueous chemicals
Membrane: UPE, nylon, PCM; Support: HDPE
Maximum operating temperature: 40 °C
Maximum forward/reverse differential pressure: 2,7 bar at 25 °C
Compatibility: PGMEA, PGME, NMP, ethyl lactate, MMP and cyclohexanone.More Product Information