Stampa…
Hand disinfectant gels, ANIOSGEL 85
Fornitore: Laboratoires Anios
1644274
DMDA1644274EA 0 EUR
DMDA1644274 DMDA1644636
Hand disinfectant gels, ANIOSGEL 85
Cura della pelle Disinfettanti per la pelle Disinfettanti per la pelle in gel
Thixotrophic hydroalcoholic gel for hygienic treatment and surgical dinfection by hand rubbing.
  • Active against bacteria, yeasts, moulds and viruses
  • Hygenic treatment: 3 ml for 30 s
  • Can be used without water
  • Colourant- and fragrant-free
Attenzione: Use biocides safely. Always read the label and product information before use.