Stampa…
Artist brushes, round
Fornitore: GORDON BRUSH MFG CO
0906-00000 0906-02000
GORD0906-00000EA 0 EUR
GORD0906-00000 GORD0906-02000
Artist brushes, round
Spazzole
This traditional round brush is made of pure red sable hair, which is softer than camel hair, pony hair and bristle.
  • Wooden handle
  • Anti-static
Ideal for applying oils, acrylics and less caustic solutions on a larger surface area.