Stampa…
SPE columns, Supra-Clean® REC18
Fornitore: PerkinElmer
N9306457 N9306491 N9306519 N9306520 N9306522
PERCN9306457EA 189 EUR
PERCN9306457 PERCN9306491 PERCN9306519 PERCN9306520 PERCN9306522
SPE columns, Supra-Clean® REC18
Colonne cromatografiche
SPE Supra-Clean® high recovery REC18 are silica based columns with 15% of carbon load. Ideal for extracting non-polar and mid-polar compounds including 100% water solvents when analysing drugs and drug metabolites in biological matrices, trace organic material in water, toxins in food.