Stampa…
Surgical gloves, latex, sterile, Garant
Fornitore: UNIGLOVES AERTZE UND KLINIKBED
1585 1575
112-1586EA 105 EUR
112-1586 112-1587
Surgical gloves, latex, sterile, Garant
Guanti
These hand-specific gloves are made of high-quality latex. Ideal for surgical use.
  • Micro-textured finish
  • Beaded cuffs
  • Hypoallergenic
  • Powdered for easier donning
Length: 270 - 280 mm
Thickness: 0,40 mm (finger), 0,35 mm (palm), 0,30 mm (cuff)
Colour: White
EN 455-1/-2/-4; AQL 1.5
Certificazioni: EN 455; AQL 1,5