Stampa…
Flame-retardant trousers, Bizweld™, BZ30
Fornitore: Portwest
BZ30BKRL BZ30NAR4XL BZ30NAR5XL BZ30NARL BZ30NARM BZ30NARS BZ30NARXL BZ30NARXS BZ30NARXXL BZ30NARXXXL BZ30NATL BZ30NATM BZ30NATS BZ30NATXL BZ30NATXXL BZ30NATXXXL
PRTWBZ30BKRLEA 45 EUR
PRTWBZ30BKRL PRTWBZ30NAR4XL PRTWBZ30NAR5XL PRTWBZ30NARL PRTWBZ30NARM PRTWBZ30NARS PRTWBZ30NARXL PRTWBZ30NARXS PRTWBZ30NARXXL PRTWBZ30NARXXXL PRTWBZ30NATL PRTWBZ30NATM PRTWBZ30NATS PRTWBZ30NATXL PRTWBZ30NATXXL PRTWBZ30NATXXXL
Flame-retardant trousers, Bizweld™, BZ30
Abbigliamento da lavoro
These trousers are made of Bizweld™ fabric consisting of 100% cotton with a flame-resistant finish. The fabric offers excellent protection for welding and all other allied industries.
  • Provide comfort and effective function at the same time
  • With ruler pocket
  • Half-elastic waistband, belt loops
Weight: 330 g/m²
Certificazioni: EN ISO 11611 (Class 1, A1+A2), EN ISO 11612 (A1+A2, B1, C1, E1, F1)