Stampa…
Chemical resistant gloves, uvex rubiflex S, NB60SZ / NB80SZ
Fornitore: UVEX
6018060 6018062 9893061 6019101 6019102 6019103
UVEX6018060EA 444 EUR
UVEX6018060 UVEX6018062 UVEX9893061 UVEX6019101 UVEX6019102 UVEX6019103
Chemical resistant gloves, uvex rubiflex S, NB60SZ / NB80SZ
Guanti
These chemical protection gloves feature a thick cotton interlock lining and a full NBR coating (nitrile rubber). This makes the uvex rubiflex S suitable for many applications in which protection against chemicals and increased mechanical protection (for example against heat) is required.
  • Good resistance to chemicals, acids, alkalis, mineral oils and solvents
  • High water vapour absorption due to the cotton lining
  • Excellent mechanical properties
  • Extremely high flexibility
  • Elasticated gauntlet end
Typical applications include: Chemical industry, sewer construction, municipal cleaning, sandblasting.
Thickness: 0,50 mm
Colour: Light green
EN 374 (JKL), EN 388 (2121)
Certificazioni: EN ISO 374-1:2016 Type B (JKOPT), EN 388:2016 (2121X). Food contact approved. OEKO-Tex Standard 100.