Stampa…
Nitrile gloves, SivoChem® 759
Fornitore: KCL by Honeywell
759.8 759.10 759.7 759.9
112-2988EA 127 EUR
112-2988 112-2986 112-2987 KCHL759.9
Nitrile gloves, SivoChem® 759
Guanti
These disposable gloves made of special nitrile offer a high level of chemical resistance and excellent comfort.
  • Non sterile, powder-free, smooth finish
  • Very good tactile sensitivity
  • With beaded cuffs
  • Naturally latex-free, free of the allergens thiuram and rosin
Ideal for use in laboratories, research environments, the chemical industry, biotechnology, medical engineering, the pharmaceutical industry and for cleaning and maintenance work.
Length: 390-410 mm
Thickness: 0.16 ± 0.03 mm
Colour: blue
EN 374; EN 388 (0000); AQL 1.5; Cat. III
Visualizza i codici
Specifiche
Modello 759 759
Ambidestro/mano specifico ambidestro ambidestro
Environmentally Preferable
Monouso/riutilizzabile monouso monouso
Lattice/senza lattice Senza lattice Senza lattice
Talcati/non talcati Non talcati Non talcati
Tipo polsino Cuff_Style-21391103|Cuff_Style-21391099 Cuff_Style-21391099|Cuff_Style-21391103
Norma EN EN ISO 374-1:2016 Type B (JKPT), EN 388:2016 (100XX) EN ISO 374-1:2016 Type B (JKPT), EN 388:2016 (100XX)