Stampa…
Dialamatic microdispensers
Fornitore: DRUMMOND
3-000-210 3-000-275 3-000-225 3-000-250
DRUM3-000-210EA 983 EUR
DRUM3-000-210 DRUM3-000-275 DRUM3-000-225 DRUM3-000-250
Dialamatic microdispensers
Pipette Pipette ripetitive
Dialamatic microdispensers with Borosilicate glass bores with stainless steel plungers (PTFE tips on all plungers 25 μl or larger).
  • Adjustable, direct reading plunger sets volume
  • Delivery scale engraved on plunger shaft
  • Fractional deliveries possible with premarked indexes
  • Volumetric accuracy of ±1% (at full volume)