Stampa…

Joining fittings, BOLA

Fornitore: Bohlender
F762-18 F762-14
BOHLF762-18EA 93 EUR
BOHLF762-18 BOHLF762-14
Joining fittings, BOLA
Raccordi e connettori
These fitting are made of PTFE, ETFE.

  • Temperature resistant from –50 to 150 °C
  • Chemical resistant
  • Pressure range up to 5 bar
  • Suitable for vacuum

One side with connecting nut with compression rings for connecting tubing or tubes and other side with laboratory screw joint for connecting tubing.
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR