Stampa…
General purpose gloves, HyFlex® 11-818
Fornitore: Ansell
11-818/10 11-818/11 11-818/6 11-818/7 11-818/8 11-818/9
112-5135EA 69.2 EUR
112-5135 112-5136 112-5131 112-5132 112-5133 112-5134
General purpose gloves, HyFlex® 11-818
Guanti
This knitted spandex/nylon liner features a foam nitrile coating on the palm. Under abrasive conditions the ultra-thin FORTIX™ nitrile foam coating lasts up to three times longer than comparable coatings, extending glove life and reducing total cost of ownership.
  • FORTIX™ coating provides enhanced dry grip and is breathable, for cooler, drier hands and less sweat
  • Seamless 18-gauge nylon-spandex liner has a barehand feel, offering breathability, comfort and enhanced fingertip sensitivity for precision tasks
  • Ansell ZONZ® Comfort Fit technology matches natural
More Product Information
Visualizza i codici
Specifiche
Modello 11-818
Environmentally Preferable
Ambidestro/mano specifico mano specifico
Monouso/riutilizzabile riutilizzabile
Materiale rivestimento interno Spandex/nylon
Finitura Foam coating on palm
Calibro 18
Tipo polsino Maglia elastica
Norma EN EN 388:2016 (3121A)