Stampa…

N-Acetyl-5-hydroxytryptamine

Fornitore: Cayman Chemical

Synonyms: N-(2-(5-Hydroxy-1H-indol-3-yl)ethyl)acetamide, N-Acetylserotonin

14535-50 14535-100 14535-250
CAYM14535-50EA 81.5 EUR
CAYM14535-50 CAYM14535-100 CAYM14535-250
N-Acetyl-5-hydroxytryptamine
N-Acetyl-5-hydroxytryptamine
Formula: C₁₂H₁₄N₂O₂
Peso molecolare: 218,3 g/mol
MDL Number: MFCD00005656
CAS Number: 1210-83-9
EINECS: 214-916-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR