Stampa…

Fmoc-Met-OH ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: N-Fmoc-L-methionine, FMOC-L-Methionine

F0296-1G F0296-25G F0296-5G
TCIAF0296-1GEA 16 EUR
TCIAF0296-1G TCIAF0296-25G TCIAF0296-5G
Fmoc-Met-OH ≥98.0% (by HPLC, titration analysis)
Fmoc-Met-OH
Formula: C₂₀H₂₁NO₄S
Peso molecolare: 371,46 g/mol
MDL Number: MFCD00037134
CAS Number: 71989-28-1
EINECS: 276-258-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f861458d923925123c25ff8d3a6fff6940950be5&vid=f861458d923925123c25ff8d3a6fff6940950be5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 140.0 to 143.0 deg-C
Specific rotation [a]20/D -27.0 to -32.0 deg (C=1, DMF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR