Stampa…

Benzo-15-crown-5 (benzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Benzo-15-crown-5

B1154-1G B1154-5G
TCIAB1154-1GEA 66 EUR
TCIAB1154-1G TCIAB1154-5G
Benzo-15-crown-5 (benzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥98.0% (by GC)
Benzo-15-crown-5 (benzo-1,4,7,10,13-pentaoxacyclopentadecane)
Formula: C₁₄H₂₀O₅
Peso molecolare: 268,31 g/mol
MDL Number: MFCD00005945
CAS Number: 14098-44-3
EINECS: 237-947-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f197b4f00e57bb6ebda2d4642b1313ffa876b781&vid=f197b4f00e57bb6ebda2d4642b1313ffa876b781&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 79.0 to 82.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR