Stampa…

4-Fluorophenethylamine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

F0829-5G F0829-25G
TCIAF0829-5GEA 22 EUR
TCIAF0829-5G TCIAF0829-25G
4-Fluorophenethylamine ≥98.0% (by GC, titration analysis)
4-Fluorophenethylamine
Formula: C₈H₁₀FN
Peso molecolare: 139,17 g/mol
Punto di ebollizione: 93°C
Flash Pt: 79°C
MDL Number: MFCD00134208
CAS Number: 1583-88-6
EINECS: 216-435-6
UN: 2735
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=009bf992c889dbff915f3fae6128467773366581&vid=009bf992c889dbff915f3fae6128467773366581&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR