2-Chlorobenzoylacetonitrile ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-(2-Chlorophenyl)-3-oxopropanenitrile
C2580-5G
C2580-25G
TCIAC2580-5GEA
103
EUR
TCIAC2580-5G
TCIAC2580-25G
2-Chlorobenzoylacetonitrile ≥97.0% (by GC)
2-Chlorobenzoylacetonitrile
Formula:
C₉H₆ClNO Peso molecolare: 179,61 g/mol Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD00051624 CAS Number: 40018-25-5 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 56.0 to 60.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...