(3-(Methylamino)propyl)trimethoxysilane ≥95.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(3-Methylaminopropyl)trimethoxysilane, Trimethoxy[3-(methylamino)propyl]silane, N-Methyl-3-(trimethoxysilyl)propylamine, Trimethoxy(3-(methylamino)propyl)silane
T2868-25G
T2868-100G
TCIAT2868-25GEA
0
EUR
TCIAT2868-25G
TCIAT2868-100G
(3-(Methylamino)propyl)trimethoxysilane ≥95.0% (by GC, titration analysis)
(3-(Methylamino)propyl)trimethoxysilane
Formula:
H₃CNHCH₂CH₂CH₂Si(OCH₃)₃ Peso molecolare: 193,32 g/mol Punto di ebollizione: 106°C Flash Pt: 82°C |
MDL Number:
MFCD00048021 CAS Number: 3069-25-8 EINECS: 221-334-5 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Purity (Nonaqueous Titration) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...