Stampa…

3-Cyclopentene-1-carboxylic acid ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: cyclopent-3-ene-1-carboxylic acid, Cyclopent-3-enecarboxylic acid

C1914-5G C1914-1G
TCIAC1914-5GEA 82 EUR
TCIAC1914-5G TCIAC1914-1G
3-Cyclopentene-1-carboxylic acid ≥97.0% (by GC, titration analysis)
3-Cyclopentene-1-carboxylic acid
Formula: C₆H₈O₂
Peso molecolare: 112,13 g/mol
MDL Number: MFCD00101764
CAS Number: 7686-77-3
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69ec66fc01decda819b6ff35127a7ea57727aed0&vid=69ec66fc01decda819b6ff35127a7ea57727aed0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR