Ethyl-3-piperidinecarboxylate ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₈H₁₅NO₂ Punto di ebollizione: 104°C Flash Pt: 90°C |
MDL Number:
MFCD00005991 CAS Number: 5006-62-2 EINECS: 225-681-3 |
|
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...