1-(4-Bromophenyl)piperidine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
B3930-1G
B3930-5G
TCIAB3930-1GEA
42
EUR
TCIAB3930-1G
TCIAB3930-5G
1-(4-Bromophenyl)piperidine ≥98.0% (by GC, titration analysis)
1-(4-Bromophenyl)piperidine
Formula:
C₁₁H₁₄BrN Peso molecolare: 240,14 g/mol |
MDL Number:
MFCD04112533 CAS Number: 22148-20-5 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 74.0 to 78.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...