trans-4-tert-Butylcyclohexanol ≥95.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
trans-4-(tert-Butyl)cyclohexanol
B1271-1G
B1271-5G
TCIAB1271-1GEA
163
EUR
TCIAB1271-1G
TCIAB1271-5G
trans-4-tert-Butylcyclohexanol ≥95.0% (by GC)
trans-4-tert-Butylcyclohexanol
Formula:
C₁₀H₂₀O Peso molecolare: 156,27 g/mol |
MDL Number:
MFCD00064952 CAS Number: 21862-63-5 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 77.0 to 81.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...