4-(Bromomethyl)-3-fluorobenzonitrile ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
B3204-1G
B3204-25G
B3204-5G
TCIAB3204-1GEA
17
EUR
TCIAB3204-1G
TCIAB3204-25G
TCIAB3204-5G
4-(Bromomethyl)-3-fluorobenzonitrile ≥98.0% (by GC)
4-(Bromomethyl)-3-fluorobenzonitrile
Formula:
C₈H₅BrFN Peso molecolare: 214,04 g/mol |
MDL Number:
MFCD07368341 CAS Number: 105942-09-4 UN: 3439 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 75.0 to 79.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...