Stampa…

Potassio acetato idrato ≥99.997% (metals basis), Puratronic®

Fornitore: Thermo Fisher Scientific

12975.06 12975.14
12975.06EA 77.7 EUR
12975.06 12975.14
Potassio acetato idrato ≥99.997% (metals basis), Puratronic®
Potassio acetato idrato

Purity: 99.997% CAS No.: 254966-93-3 Molecular Formula: CH3CO2K.xH2O Formula Weight: 98.15(anhy) Physical Form: Crystalline Appearance: White MDL No.: MFCD00012458

Formula: C₂H₃KO₂·H₂O
Peso molecolare: 98.15 g/mol (anhy)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00012458
CAS Number: 254966-93-3
EINECS: 204-822-2

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR