N,N'-Dimethyl-1,6-diaminohexane ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
N1,N6-Dimethylhexane-1,6-diamine
D1298-10ML
D1298-25ML
TCIAD1298-10MLEA
121
EUR
TCIAD1298-10ML
TCIAD1298-25ML
N,N'-Dimethyl-1,6-diaminohexane ≥97.0% (by GC)
N,N'-Dimethyl-1,6-diaminohexane
Formula:
C₈H₂₀N₂ Peso molecolare: 144,2578 g/mol Punto di ebollizione: 205°C Flash Pt: 112°C |
MDL Number:
MFCD00008293 CAS Number: 13093-04-4 EINECS: 236-005-1 UN: 2735 ADR: 8,III |
Risultati del test di specificazione
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...