Stampa…

D-Phenylalaninol ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: D-Penylalaninol, D(+)-2-Amino-3-phenyl-1-propanol

P1289-25G P1289-5G
TCIAP1289-25GEA 255 EUR
TCIAP1289-25G TCIAP1289-5G
D-Phenylalaninol ≥98.0% (by GC, titration analysis)
D-Phenylalaninol
Formula: C₉H₁₃NO
Peso molecolare: 151,2 g/mol
Punto di ebollizione: 122°C
MDL Number: MFCD00064298
CAS Number: 5267-64-1
EINECS: 226-086-1
UN: 3259
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3503cd3ba732f3940414439ff9dc51594d54e7f4&vid=3503cd3ba732f3940414439ff9dc51594d54e7f4&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 90.0 to 94.0 deg-C
Specific rotation [a]20/D +22.0 to +25.0 deg (C=1.2, 1mol/L HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR