Stampa…

2-(Hydroxymethyl)-1,4-benzodioxane ≥98.0% (by GC)

Fornitore: TCI EUROPE

H0801-25G H0801-500G
TCIAH0801-25GEA 39 EUR
TCIAH0801-25G TCIAH0801-500G
2-(Hydroxymethyl)-1,4-benzodioxane ≥98.0% (by GC)
2-(Hydroxymethyl)-1,4-benzodioxane
Formula: C₉H₁₀O₃
Peso molecolare: 166,18 g/mol
Punto di ebollizione: 286°C
MDL Number: MFCD00006822
CAS Number: 3663-82-9
EINECS: 222-921-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bde9961e31549218ce204bbb41043ae784181e70&vid=bde9961e31549218ce204bbb41043ae784181e70&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 86.0 to 90.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR