Stampa…

4-Methyloctanoic acid ≥98%

Fornitore: Thermo Fisher Scientific
Pericolo

Synonyms: 4-methylcaprylic acid, (R,S)-(+,-)-4-Methyloctanoic acid

B22616.06 B22616.14
B22616.06EA 46.7 EUR
B22616.06 B22616.14
4-Methyloctanoic acid ≥98%
4-Methyloctanoic acid

Purity: 98% CAS No.: 54947-74-9 Molecular Formula: C9H18O2 Formula Weight: 158.24 Flash Point: 110°(230°F) Refractive Index: 1.4335 Boiling Point: 272° Beilstein Reference: 1755426 UN No.: 3265 MDL No.: MFCD00051938

Formula: C₉H₁₈O₂
Peso molecolare: 158.24 g/mol
Punto di ebollizione: 272 °C
Densità: 0.910
Flash Pt: >110 °C (230 °F)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00051938
CAS Number: 54947-74-9
EINECS: 259-404-2
ADR: 8 III

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR