Stampa…

4,4'-Difluorobenzhydryl chloride ≥97.0% (by GC)

Fornitore: TCI
Pericolo

Synonyms: Chloro bis-(4-fluorophenyl)methane, 4,4-Difluorodiphenylmethylchloride

D4138-25G D4138-5G
TCIAD4138-25GEA 98 EUR
TCIAD4138-25G TCIAD4138-5G
4,4'-Difluorobenzhydryl chloride ≥97.0% (by GC)
4,4'-Difluorobenzhydryl chloride
Formula: C₁₃H₉ClF₂
Punto di ebollizione: 112°C
MDL Number: MFCD00044329
CAS Number: 27064-94-4
EINECS: 248-201-4
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07645cb35c22564f4809219a0f331dcfa45008bb&vid=07645cb35c22564f4809219a0f331dcfa45008bb&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR