Stampa…

2,5-Bis(4-aminophenyl)-1,3,4-oxadiazole ≥98.0% (by HPLC)

Fornitore: TCI
Attenzione

B2169-1G B2169-5G
TCIAB2169-1GEA 50 EUR
TCIAB2169-1G TCIAB2169-5G
2,5-Bis(4-aminophenyl)-1,3,4-oxadiazole ≥98.0% (by HPLC)
2,5-Bis(4-aminophenyl)-1,3,4-oxadiazole
Formula: C₁₄H₁₂N₄O
MDL Number: MFCD00042667
CAS Number: 2425-95-8
EINECS: 219-373-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9d84beacf4bc1bd4b01c5b6e23323419395bea9&vid=b9d84beacf4bc1bd4b01c5b6e23323419395bea9&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 96.0 % (calcd.on anh.substance)
Water max. 7.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR