2,6-Bis(bromomethyl)pyridine ≥99.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
B1936-1G
B1936-5G
TCIAB1936-1GEA
59
EUR
TCIAB1936-1G
TCIAB1936-5G
2,6-Bis(bromomethyl)pyridine ≥99.0% (by GC, titration analysis)
2,6-Bis(bromomethyl)pyridine
Formula:
C₇H₇Br₂N Peso molecolare: 264,95 g/mol |
MDL Number:
MFCD00191795 CAS Number: 7703-74-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystaline |
Purity (GC) | min. 99.0 % |
Purity (Nonaqueous Titration) | min. 99.0 % |
Melting point | 85.0 to 89.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...