4-(4-Chlorobenzoyl)pyridine ≥99%
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
Purity: 99+%
CAS No.: 14548-48-2
Molecular Formula: C12H8ClNO
Formula Weight: 217.66
Melting Point: 107-109°
MDL No.: MFCD00006431
Formula:
C₁₂H₈ClNO Punto di fusione: 107…109 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00006431 CAS Number: 14548-48-2 EINECS: 238-587-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...