2-Acetyl-4-methylpyridine ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₈H₉NO Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD01863638 CAS Number: 59576-26-0 |
|
Risultati del test di specificazione
Appearance | White to Gray to Brown powder to lump |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...