Stampa…
3-Acetyl-5-bromopyridine ≥97.0% (by GC)
Fornitore: TCI EUROPE
Attenzione
A2376-1G A2376-5G
TCIAA2376-1GEA 25 EUR
TCIAA2376-1G TCIAA2376-5G
3-Acetyl-5-bromopyridine ≥97.0% (by GC)
3-Acetyl-5-bromopyridine
Formula: C₇H₆BrNO
Peso molecolare: 200,03 g/mol
MDL Number: MFCD03086033
CAS Number: 38940-62-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5ea8849be8fbce258fd73fedf124c94281fbadc&vid=f5ea8849be8fbce258fd73fedf124c94281fbadc&prid=web&clid=SAPDAM
Visualizza i codici
Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 90.0 to 94.0 deg-C