Silver(II) picolinate ≥97.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
S0815-1G
S0815-5G
TCIAS0815-1GEA
74
EUR
TCIAS0815-1G
TCIAS0815-5G
Silver(II) picolinate ≥97.0% (by titrimetric analysis)
Silver(II) picolinate
Formula:
C₁₂H₈AgN₂O₄ Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD15072194 CAS Number: 14783-00-7 |
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
Purity (Neutralization titration) | min. 97.0 % |
Purity (Precipitation Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...