Stampa…

3-Carbamoylpicolinic acid ≥97.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

C1508-1G C1508-100MG
TCIAC1508-1GEA 486 EUR
TCIAC1508-1G TCIAC1508-100MG
3-Carbamoylpicolinic acid ≥97.0% (by titrimetric analysis)
3-Carbamoylpicolinic acid
Formula: C₇H₆N₂O₃
MDL Number: MFCD00661289
CAS Number: 4733-65-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2495918ca134e6a37f77833ab0e5f071d7f05f75&vid=2495918ca134e6a37f77833ab0e5f071d7f05f75&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR