Stampa…

Calmagite metal indicator for Ca, Mg etc.

Fornitore: TCI
Attenzione

C0008-1G C0008-5G
TCIAC0008-1GEA 46 EUR
TCIAC0008-1G TCIAC0008-5G
Calmagite metal indicator for Ca, Mg etc.
Calmagite
Formula: C₁₇H₁₄N₂O₅S
Punto di fusione: 330°C
MDL Number: MFCD00011656
CAS Number: 3147-14-6
EINECS: 221-563-0
Merck Index: 14,01718
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f507ac2142bef470f267714630199a51218d1f27&vid=07faff2ff13319fcd1c4c63b77b49046ebd350d8&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Red to Dark blue to Black powder to crystal
Sensitiveness Turn Red with Calcium carbonate and turn Blue by adding EDTA 2Na


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR