Stampa…

Potassium(iodomethyl)trifluoroborate ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

Synonyms: Potassium trifluoro(iodomethyl)borate

P1808-1G P1808-5G
TCIAP1808-1GEA 64 EUR
TCIAP1808-1G TCIAP1808-5G
Potassium(iodomethyl)trifluoroborate ≥98.0% (by titrimetric analysis)
Potassium(iodomethyl)trifluoroborate
Formula: CH₂BF₃IK
Punto di fusione: 242°C
MDL Number: MFCD11112187
CAS Number: 888711-47-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=35a14a25dd597021913c1f789d6124df253cfc3d&vid=35a14a25dd597021913c1f789d6124df253cfc3d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Potassium iodate Method) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR