Stampa…

trans-4-(Hydroxymethyl)cyclohexanol ≥98.0% (by GC)

Fornitore: TCI EUROPE

H1258-1G H1258-5G
TCIAH1258-1GEA 135 EUR
TCIAH1258-1G TCIAH1258-5G
trans-4-(Hydroxymethyl)cyclohexanol ≥98.0% (by GC)
trans-4-(Hydroxymethyl)cyclohexanol
Formula: C₇H₁₄O₂
Peso molecolare: 130,1900 g/mol
MDL Number: MFCD11036248
CAS Number: 3685-27-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcc274283fc543485f9721df37a80421a7fd09c8&vid=dcc274283fc543485f9721df37a80421a7fd09c8&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 102.0 to 106.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR