Stampa…

4-(4-Aminophenyl)benzonitrile ≥95.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Amino-[1,1-biphenyl]-4-carbonitrile, 4'-Aminobiphenyl-4-carbonitrile

A1249-1G A1249-5G
TCIAA1249-1GEA 95 EUR
TCIAA1249-1G TCIAA1249-5G
4-(4-Aminophenyl)benzonitrile ≥95.0% (by GC)
4-(4-Aminophenyl)benzonitrile
Formula: C₁₃H₁₀N₂
Peso molecolare: 194,2319 g/mol
MDL Number: MFCD00191367
CAS Number: 4854-84-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f164f2617c57134b86971054d9939f01316a1a3&vid=5f164f2617c57134b86971054d9939f01316a1a3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Amber powder to crystal
Purity (GC) min. 95.0 %
Melting point 182.0 to 185.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR