Stampa…

(±)-Cbz-α-phosphonoglycine trimethyl ester ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE

Synonyms: N-Benzyloxycarbonyl-2-phosphonoglycine trimethyl ester, (±)-Benzyloxycarbonyl-α-phosphonoglycine trimethyl ester, (±)-Methyl 2-benzyloxycarbonylamino-2-(dimethoxyphosphinyl)acetate, Methyl 2-{[(benzyloxy)carbonyl]amino}-2-(dimethoxyphosphoryl)acetate, Methyl 2-(((benzyloxy)carbonyl)amino)-2-(dimethoxyphosphoryl)acetate

C2440-1G C2440-5G
TCIAC2440-1GEA 27 EUR
TCIAC2440-1G TCIAC2440-5G
(±)-Cbz-α-phosphonoglycine trimethyl ester ≥98.0% (by HPLC, total nitrogen)
(±)-Cbz-α-phosphonoglycine trimethyl ester
Formula: C₁₃H₁₈NO₇P
Peso molecolare: 331,27 g/mol
MDL Number: MFCD00043304
CAS Number: 88568-95-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb681feb3b3cdee8fd51f33326fffbe99b2382b3&vid=eb681feb3b3cdee8fd51f33326fffbe99b2382b3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 78.0 to 82.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR