N-Cbz-L-phenylalaninol ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
Z-Phenylalaninol, (S)-2-(Cbz-amino)-3-phenyl-1-propanol, N-Benzyloxycarbonyl-L-phenylalaninol
C1610-1G
C1610-5G
TCIAC1610-1GEA
21
EUR
TCIAC1610-1G
TCIAC1610-5G
N-Cbz-L-phenylalaninol ≥98.0% (by HPLC)
N-Cbz-L-phenylalaninol
Formula:
C₁₇H₁₉NO₃ Peso molecolare: 285,3 g/mol |
MDL Number:
MFCD00191138 CAS Number: 6372-14-1 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Optical purity (LC) | min. 98.0 ee% |
Melting point | 91.0 to 95.0 deg-C |
Specific rotation [a]20/D | -40.0 to -44.0 deg (C=2, MeOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...