Stampa…

N-Cbz-D-phenylalaninol ≥97.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: N-Benzyloxycarbonyl-D-phenylalaninol, Z-D-Phenylalaninol, Cbz-D-Phenylalaninol

C1609-1G C1609-5G
TCIAC1609-1GEA 21 EUR
TCIAC1609-1G TCIAC1609-5G
N-Cbz-D-phenylalaninol ≥97.0% (by HPLC)
N-Cbz-D-phenylalaninol
Formula: C₁₇H₁₉NO₃
Peso molecolare: 285,3 g/mol
MDL Number: MFCD00191193
CAS Number: 58917-85-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d653d2861281a9d0a6e4625e2460d3197d60dd6d&vid=d653d2861281a9d0a6e4625e2460d3197d60dd6d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Optical purity (LC) min. 98.0 ee%
Melting point 91.0 to 95.0 deg-C
Specific rotation [a]20/D +40.0 to +44.0 deg (C=2, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR