Stampa…

Methyl-4-(hydroxymethyl)benzoate ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: Methyl 4-(hydroxymethyl)benzoate

H0640-25G H0640-5G
TCIAH0640-25GEA 105 EUR
TCIAH0640-25G TCIAH0640-5G
Methyl-4-(hydroxymethyl)benzoate ≥98.0% (by titrimetric analysis)
Methyl-4-(hydroxymethyl)benzoate
Formula: C₉H₁₀O₃
Peso molecolare: 166,18 g/mol
Punto di ebollizione: 157°C
MDL Number: MFCD00009984
CAS Number: 6908-41-4
EINECS: 230-017-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bef132a1a315f9e2601fae6e70942f6746273e46&vid=bef132a1a315f9e2601fae6e70942f6746273e46&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Ester Value) min. 98.0 %
Melting point 48.0 to 52.0 deg-C
Solubility in Methanol within almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR