Stampa…

3-Amino-4-methylbenzamide ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

A2367-25G A2367-5G
TCIAA2367-25GEA 114 EUR
TCIAA2367-25G TCIAA2367-5G
3-Amino-4-methylbenzamide ≥98.0% (by HPLC, titration analysis)
3-Amino-4-methylbenzamide
Formula: C₈H₁₀N₂O
Peso molecolare: 150,18 g/mol
MDL Number: MFCD00035936
CAS Number: 19406-86-1
EINECS: 243-039-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=704a8c795ec8810f86ec282bdfd50cfffe91dd08&vid=704a8c795ec8810f86ec282bdfd50cfffe91dd08&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 129.0 to 133.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR