Stampa…

N-Boc-DL-phenylalaninol ≥96.0% (by GC)

Fornitore: TCI

Synonyms: Boc-DL-Phenylalaninol, tert-Butyl (1-hydroxy-3-phenylpropan-2-yl)carbamate

B3269-1G B3269-5G
TCIAB3269-1GEA 38 EUR
TCIAB3269-1G TCIAB3269-5G
N-Boc-DL-phenylalaninol ≥96.0% (by GC)
N-Boc-DL-phenylalaninol
Formula: C₁₄H₂₁NO₃
MDL Number: MFCD01457313
CAS Number: 145149-48-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98edd944575c51038d9609bf86568a1d30fcfcd0&vid=98edd944575c51038d9609bf86568a1d30fcfcd0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Melting point 85.0 to 90.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR