Stampa…
(1H-Imidazol-4-yl)methanol hydrochloride 99%
Fornitore: Thermo Scientific
Attenzione
Synonyms: 4-Imidazolemethanol hydrochloride, 4(5)-Imidazolemethanol hydrochloride, 4-(Hydroxymethyl)imidazole hydrochloride
204870010 204870050
ACRO204870010EA 50.3 EUR
ACRO204870010 ACRO204870050
(1H-Imidazol-4-yl)methanol hydrochloride 99%
(1H-Imidazol-4-yl)methanol hydrochloride
Formula: C₄H₇ClN₂O
Peso molecolare: 134,57 g/mol
Punto di fusione: 110…112 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00012697
CAS Number: 32673-41-9
EINECS: 251-150-0
Visualizza i codici
Appearance Light yellow to beige to light brown Crystalline powder and/or chunks
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %