Stampa…

1,4-Bis(4-aminophenoxy)benzene ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

B1243-25G B1243-5G
TCIAB1243-25GEA 86 EUR
TCIAB1243-25G TCIAB1243-5G
1,4-Bis(4-aminophenoxy)benzene ≥98.0% (by GC, titration analysis)
1,4-Bis(4-aminophenoxy)benzene
Formula: C₁₈H₁₆N₂O₂
MDL Number: MFCD00039153
CAS Number: 3491-12-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f07ec474c15ca7dbda76be727268686b77d069c9&vid=f07ec474c15ca7dbda76be727268686b77d069c9&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 173.0 to 177.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR