Stampa…

2-Aminophenol-4-sulphonamide ≥97.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Amino-4-hydroxybenzenesulphonamide

A1031-25G A1031-500G
TCIAA1031-25GEA 18 EUR
TCIAA1031-25G TCIAA1031-500G
2-Aminophenol-4-sulphonamide ≥97.0% (by HPLC, titration analysis)
2-Aminophenol-4-sulphonamide
Formula: C₆H₈N₂O₃S
Peso molecolare: 188,2 g/mol
MDL Number: MFCD00035782
CAS Number: 98-32-8
EINECS: 202-657-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0534a6cc6d3fe1bf39a9f51effeceaaf5d819ae1&vid=0534a6cc6d3fe1bf39a9f51effeceaaf5d819ae1&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Gray to Brown powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR