Stampa…

N-Boc-L-valine ≥99.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: Boc-Val-OH, BOC-L-Valine

B1333-25G B1333-5G
TCIAB1333-25GEA 42 EUR
TCIAB1333-25G TCIAB1333-5G
N-Boc-L-valine ≥99.0% (by titrimetric analysis)
N-Boc-L-valine
Formula: C₁₀H₁₉NO₄
Peso molecolare: 217,3 g/mol
MDL Number: MFCD00065605
CAS Number: 13734-41-3
EINECS: 237-307-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae807aa94496dce674c7d40c7fd85d8c5bb93fd8&vid=ae807aa94496dce674c7d40c7fd85d8c5bb93fd8&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 99.0 %
Melting point 78.0 to 81.0 deg-C
Specific rotation [a]20/D -6.2 to -6.7 deg (C=1, AcOH)
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR