Stampa…

1,2-Ethanedisulphonic acid disodium salt ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Sodium ethane-1,2-disulphonate

E0031-25G E0031-5G
TCIAE0031-25GEA 107 EUR
TCIAE0031-25G TCIAE0031-5G
1,2-Ethanedisulphonic acid disodium salt ≥98.0% (by titrimetric analysis)
1,2-Ethanedisulphonic acid disodium salt
Formula: C₂H₄Na₂O₆S₂
Peso molecolare: 234,1600 g/mol
MDL Number: MFCD00024998
CAS Number: 5325-43-9
EINECS: 226-198-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dec8f46ef5bc5f5dad34c7dc5dc4ebba09d69c94&vid=dec8f46ef5bc5f5dad34c7dc5dc4ebba09d69c94&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystaline
Purity (Ion exchange titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR